Merck
  • Acute frataxin knockdown in induced pluripotent stem cell-derived cardiomyocytes activates a type I interferon response.
Acute frataxin knockdown in induced pluripotent stem cell-derived cardiomyocytes activates a type I interferon response.
Disease models & mechanisms (2022-09-16)
M Grazia Cotticelli, Shujuan Xia, Rachel Truitt, Nicolai M Doliba, Andrea V Rozo, John W Tobias, Taehee Lee, Justin Chen, Jill S Napierala, Marek Napierala, Wenli Yang, Robert B Wilson
ABSTRACT
Friedreich ataxia, the most common hereditary ataxia, is a neuro- and cardio-degenerative disorder caused, in most cases, by decreased expression of the mitochondrial protein frataxin. Cardiomyopathy is the leading cause of premature death. Frataxin functions in the biogenesis of iron-sulfur clusters, which are prosthetic groups that are found in proteins involved in many biological processes. To study the changes associated with decreased frataxin in human cardiomyocytes, we developed a novel isogenic model by acutely knocking down frataxin, post-differentiation, in cardiomyocytes derived from induced pluripotent stem cells (iPSCs). Transcriptome analysis of four biological replicates identified severe mitochondrial dysfunction and a type I interferon response as the pathways most affected by frataxin knockdown. We confirmed that, in iPSC-derived cardiomyocytes, loss of frataxin leads to mitochondrial dysfunction. The type I interferon response was activated in multiple cell types following acute frataxin knockdown and was caused, at least in part, by release of mitochondrial DNA into the cytosol, activating the cGAS-STING sensor pathway.
Sigma-Aldrich
RU.521, ≥98% (HPLC)
Sigma-Aldrich
Anti-dsRNA Antibody, clone rJ2, culture supernatant, clone rJ2, from mouse