Merck
  • Infection of SARS-CoV-2 causes severe pathological changes in mouse testis.
Infection of SARS-CoV-2 causes severe pathological changes in mouse testis.
Journal of genetics and genomics = Yi chuan xue bao (2022-12-10)
Min Chen, Shihua Li, Shujun Liu, Yuhang Zhang, Xiuhong Cui, Limin Lv, Bowen Liu, Aihua Zheng, Qihui Wang, Shuguang Duo, Fei Gao
ABSTRACT
Coronavirus disease 2019 (COVID-19), caused by severe acute respiratory syndrome coronavirus 2 (SARS-CoV-2), has affected more than 600 million people worldwide. Several organs including lung, intestine, and brain are infected by SARS-CoV-2. It has been reported that SARS-CoV-2 receptor angiotensin-converting enzyme-2 (ACE2) is expressed in human testis. However, whether testis is also affected by SARS-CoV-2 is still unclear. In this study, we generate a human ACE2 (hACE2) transgenic mouse model in which the expression of hACE2 gene is regulated by hACE2 promoter. Sertoli and Leydig cells from hACE2 transgenic mice can be infected by SARS-CoV-2 pseudovirus in vitro, and severe pathological changes are observed after injecting the SARS-CoV-2 pseudovirus into the seminiferous tubules. Further studies reveal that Sertoli and Leydig cells from hACE2 transgenic mice are also infected by authentic SARS-CoV-2 virus in vitro. After testis interstitium injection, authentic SARS-CoV-2 viruses are first disseminated to the interstitial cells, and then detected inside the seminiferous tubules which in turn cause germ cell loss and disruption of seminiferous tubules. Our study demonstrates that testis is most likely a target of SARS-CoV-2 virus. Attention should be paid to the reproductive function in SARS-CoV-2 patients.
Sigma-Aldrich
Monoclonal Anti-SOX9 antibody produced in mouse, Prestige Antibodies® Powered by Atlas Antibodies, clone CL0639, purified immunoglobulin, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-Sox9 Antibody, Chemicon®, from rabbit