Merck
  • Correlation of Beta2-Glycoprotein I With Tumor Prognosis in Breast Cancer Patients.
Correlation of Beta2-Glycoprotein I With Tumor Prognosis in Breast Cancer Patients.
Anticancer research (2023-07-28)
Yi-Chen Lee, Chung-Tao Tang, Jung-Yu Kan, Chih-Po Chiang, Huang-Tse Li, Jie-Ning Sung, Wen-Chin Chiu
ABSTRACT
Beta2-glycoprotein I (β2-GPI) is a plasma glycoprotein, which has been implicated in a variety of physiological functions. However, the connection between β2-GPI and breast cancer is mostly unknown. Breast cancer is a malignant tumor that severely impairs women's health worldwide. The aim of the study was to investigate the role of β2-GPI in tumor cells of breast cancer patients and its correlation with tumor prognosis. A total of 125 female patients diagnosed with breast cancer were enrolled in the study. The expression of β2-GPI in resected breast tissues was determined by immunohistochemistry (IHC) and correlated with clinicopathological variables by the Chi-squared test. The prognostic value of β2-GPI for overall survival (OS) and disease-free survival (DFS) was determined by Kaplan-Meier estimates and the significance of differences was evaluated by the log-rank test. β2-GPI staining was predominantly observed in tumor cells of breast cancer patients and significantly correlated with tumor stage and lymph node metastasis of breast cancer. High β2-GPI expression was significantly correlated with better OS and DFS. Moreover, DFS was found to be significantly better in patients with higher β2-GPI expression, especially those in the early tumor stage groups. High β2-GPI expression levels in tumor cells of breast cancer patients were independent factors predicting a better OS and DFS. β2-GPI activation in high-risk patients may be a potential strategy for reducing breast cancer progression.
Sigma-Aldrich
Anti-APOH antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution