Merck
  • A "multi-omics" analysis of blood-brain barrier and synaptic dysfunction in APOE4 mice.
A "multi-omics" analysis of blood-brain barrier and synaptic dysfunction in APOE4 mice.
The Journal of experimental medicine (2022-08-31)
Giuseppe Barisano, Kassandra Kisler, Brent Wilkinson, Angeliki Maria Nikolakopoulou, Abhay P Sagare, Yaoming Wang, William Gilliam, Mikko T Huuskonen, Shu-Ting Hung, Justin K Ichida, Fan Gao, Marcelo P Coba, Berislav V Zlokovic
ABSTRACT
Apolipoprotein E4 (APOE4), the main susceptibility gene for Alzheimer's disease, leads to blood-brain barrier (BBB) breakdown in humans and mice. Remarkably, BBB dysfunction predicts cognitive decline and precedes synaptic deficits in APOE4 human carriers. How APOE4 affects BBB and synaptic function at a molecular level, however, remains elusive. Using single-nucleus RNA-sequencing and phosphoproteome and proteome analysis, we show that APOE4 compared with APOE3 leads to an early disruption of the BBB transcriptome in 2-3-mo-old APOE4 knock-in mice, followed by dysregulation in protein signaling networks controlling cell junctions, cytoskeleton, clathrin-mediated transport, and translation in brain endothelium, as well as transcription and RNA splicing suggestive of DNA damage in pericytes. Changes in BBB signaling mechanisms paralleled an early, progressive BBB breakdown and loss of pericytes, which preceded postsynaptic interactome disruption and behavioral deficits that developed 2-5 mo later. Thus, dysregulated signaling mechanisms in endothelium and pericytes in APOE4 mice reflect a molecular signature of a progressive BBB failure preceding changes in synaptic function and behavior.
Sigma-Aldrich
Anti-Glucose Transporter GLUT-1 Antibody, CT, Chemicon®, from rabbit
Sigma-Aldrich
Anti-NeuN Antibody (rabbit), from rabbit, purified by affinity chromatography