Merck
  • Optimization of differential filtration-based mitochondrial isolation for mitochondrial transplant to cerebral organoids.
Optimization of differential filtration-based mitochondrial isolation for mitochondrial transplant to cerebral organoids.
Stem cell research & therapy (2023-08-15)
David F Bodenstein, Pavel Powlowski, Kassandra A Zachos, Dana El Soufi El Sabbagh, Hyunjin Jeong, Liliana Attisano, Landon Edgar, Douglas C Wallace, Ana Cristina Andreazza
ABSTRACT
Mitochondrial dysfunction is involved in several diseases ranging from genetic mitochondrial disorders to chronic metabolic diseases. An emerging approach to potentially treat mitochondrial dysfunction is the transplantation of autologous live mitochondria to promote cell regeneration. We tested the differential filtration-based mitochondrial isolation protocol established by the McCully laboratory for use in cellular models but found whole cell contaminants in the mitochondrial isolate. Therefore, we explored alternative types of 5-μm filters (filters A and B) for isolation of mitochondria from multiple cell lines including HEK293 cells and induced pluripotent stem cells (iPSCs). MitoTracker™ staining combined with flow cytometry was used to quantify the concentration of viable mitochondria. A proof-of-principle mitochondrial transplant was performed using mitoDsRed2-tagged mitochondria into a H9-derived cerebral organoid. We found that filter B provided the highest quality mitochondria as compared to the 5-μm filter used in the original protocol. Using this method, mitochondria were also successfully isolated from induced pluripotent stem cells. To test for viability, mitoDsRed2-tagged mitochondria were isolated and transplanted into H9-derived cerebral organoids and observed that mitochondria were engulfed as indicated by immunofluorescent co-localization of TOMM20 and MAP2. Thus, use of filter B in a differential filtration approach is ideal for isolating pure and viable mitochondria from cells, allowing us to begin evaluating long-term integration and safety of mitochondrial transplant using cellular sources.
Sigma-Aldrich
Anti-E1A (Ad2/Ad5) Antibody, clone M73, clone M73, Upstate®, from mouse
Pricing and availability is not currently available.