Merck
  • A protocol for concurrent high-quality immunohistochemical and biochemical analyses in adult mouse central nervous system.
A protocol for concurrent high-quality immunohistochemical and biochemical analyses in adult mouse central nervous system.
The European journal of neuroscience (2013-12-12)
Tina Notter, Patrizia Panzanelli, Sandra Pfister, Dennis Mircsof, Jean-Marc Fritschy
ABSTRACT
Biochemical analysis of central nervous system proteins and nucleic acids requires fresh-tissue homogenates, whereas immunohistochemistry usually is performed in sections prepared from perfusion-fixed tissue. Post-mortem immersion-fixation is possible, but largely impairs morphological preservation and protein antigenicity. Here, we present a simple, fast and versatile protocol allowing concurrent biochemical and immunohistochemical analysis, including pre-embedding immunoelectron microscopy, using tissue from the same animal. The protocol includes a brief transcardiac perfusion with ice-cold, oxygenated and glucose-supplemented artificial cerebrospinal fluid to maintain brain tissue alive, prior to isolation of regions of interest, followed by homogenisation for biochemistry or immersion-fixation for immunohistochemistry. We provide several examples demonstrating that this protocol allows optimal biochemical and morphological analysis, characterised with optimal sensitivity and preservation of tissue structure, along with a reduction of artefacts typically seen in perfusion-fixed tissue. This protocol should find widespread applications for combining analytical methods in tissue from the same animal, thereby reducing the number of mice required for a given experiment.
Sigma-Aldrich
Anti-APP A4 Antibody, a.a. 66-81 of APP {NT}, clone 22C11, clone 22C11, Chemicon®, from mouse
Sigma-Aldrich
Anti-Tyrosine Hydroxylase Antibody, Chemicon®, from rabbit
Sigma-Aldrich
Anti-Reelin Antibody, a.a. 164-496 mreelin, clone G10, clone G10, Chemicon®, from mouse