Merck
  • Enkurin domain containing 1 (ENKD1) regulates the proliferation, migration and invasion of non-small cell lung cancer cells.
Enkurin domain containing 1 (ENKD1) regulates the proliferation, migration and invasion of non-small cell lung cancer cells.
Asia-Pacific journal of clinical oncology (2021-03-17)
Ting Song, Peng Zhou, Chunjiao Sun, Na He, Haixia Li, Jie Ran, Jun Zhou, Yue Wu, Min Liu
ABSTRACT
Non-small cell lung cancer (NSCLC) is the most common cause of cancer mortality worldwide. NSCLC has an aggressive phenotype and poor prognosis, and is quite heterogeneous without effective and specific targeted therapies. Therefore, exploring new tumor markers and drug targets for NSCLC is crucial towards individualized treatment. Here, we demonstrate that enkurin domain containing 1 (ENKD1), a protein with unknown structure and function, is significantly downregulated in NSCLC tumor tissues compared with their non-tumor counterparts. We also show that ENKD1 expression is decreased in NSCLC cells compared to normal human lung epithelial cells. EdU incorporation, wound healing, and transwell invasion assays reveal that ENKD1 regulates the proliferation, migration, and invasion of NSCLC cells. Collectively, these results suggest that ENKD1 plays an important role in NSCLC progression and that ENKD1 is a tumor marker and a potential molecular drug target for the treatment of NSCLC patients.
Sigma-Aldrich
Anti-ENKD1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution