- High-level production of a humanized immunoRNase fusion protein from stably transfected myeloma cells.
High-level production of a humanized immunoRNase fusion protein from stably transfected myeloma cells.
Methods in molecular biology (Clifton, N.J.) (2009-03-03)
Jürgen Krauss, Evelyn Exner, Athanasios Mavratzas, Siegfried Seeber, Michaela A E Arndt
PMID19252845
ABSTRACT
ImmunoRNases represent a highly attractive alternative to conventional immunotoxins for cancer therapy. Quantitative production of immunoRNases in appropriate expression systems, however, remains a major challenge for further clinical development of these novel compounds. Here we describe a method for high-level production and purification of a fully functional immunoRNase fusion protein from supernatants of stably transfected mammalian cells.