Merck
  • Metastatic Osteolytic Angioleiomyosarcoma Induced by a Foreign Body in a Kangaroo (Macropus giganteus).
Metastatic Osteolytic Angioleiomyosarcoma Induced by a Foreign Body in a Kangaroo (Macropus giganteus).
Australian veterinary journal (2019-07-10)
L Setyo, M Sabater, A Young, B H Rickman
ABSTRACT
An adult female Eastern Grey kangaroo (Macropus giganteus) was examined for lameness due to a mass located at the right tibiotarsal joint. Radiographs revealed a metal arrowhead-shaped foreign body within the mass alongside a pathologic fracture. Grossly, at necropsy, there was a multilobulated and haemorrhagic mass with intralesional metal fragments, associated osteolysis of the tibiotarsus and pulmonary metastatic nodules. Histologically the neoplasm was composed of plump elongated and fusiform cells that often formed concentrically around small vessels. The pulmonary nodules were composed of similar neoplastic cells, necrosis and haemorrhage. Neoplastic cells were positive immunohistochemically for vimentin and smooth muscle actin. The histological features, immunohistochemical profiles and behaviour of this tumour support a diagnosis of a primary angioleiomyosarcoma with lung metastasis associated with a metal foreign body.
Sigma-Aldrich
Anti-PRX antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Pricing and availability is not currently available.