Merck
  • Peptide-based fluorescence biosensors for detection/measurement of nanoparticles.
Peptide-based fluorescence biosensors for detection/measurement of nanoparticles.
Analytical and bioanalytical chemistry (2016-12-03)
Oluyemi Akinloye, Ramanarayan Krishnamurthy, David Wishart, Greg G Goss
ABSTRACT
The ability to detect and quantify nanoparticles is essential but there is currently no simple, sensitive, and rapid method for the detection of nanomaterials. We have developed a novel peptide-based fluorescence-based biosensor for detection and measurement of negatively charged engineered nanoparticles (ENPs). A peptide biosensor (seven lysine residues linked to a cysteine through a three glycine residue linker) with attached fluorescent probes-fluorescein-5-maleimide (F5M) and tetramethylrhodamine-5-maleimide (TMR5M)-was constructed. The fluorescent probes allow close monitoring of the molecular interaction of the labeled peptide with ENPs. The ENP-peptide interaction induces the formation of agglomerates that can be detected and measured by changes in the fluorescence intensities of the labeled peptides or/and by differential light scattering. The relative fluorescence intensities of F5M and TMR5M decreased in a concentration-dependent manner on interaction with various types of negatively charged ENPs (ZnO, Fe3O4, CeO, and single-walled carbon nanotubes). Differential light scattering measurements also showed increases in the hydrodynamic size of the complex. The interactions were not affected by the pH of aqueous media, where humic acid (1 μg/mL) quenched the fluorescence intensity of F5M by approximately 25 %, whereas that of TMR5M was completely quenched. Interference by humic acid at lower concentrations was less prevalent. This novel method is a simple, rapid, and inexpensive in situ assay that shows promise as a primary-level testing technique for detection of ENPs in environmental samples. Graphical Abstract Detection of nanomaterials in aqueous solutions using fluorescently-labeled designer peptides.
Sigma-Aldrich
N-(5-Fluoresceinyl)maleimide, suitable for fluorescence, ≥90% (HPLC), BioReagent