Merck
  • Fluorescence microscopy of rat embryo sections stained with haematoxylin-eosin and Masson's trichrome method.
Fluorescence microscopy of rat embryo sections stained with haematoxylin-eosin and Masson's trichrome method.
Journal of microscopy (1998-09-02)
J M Apgar, A Juarranz, J Espada, A Villanueva, M Cañete, J C Stockert
ABSTRACT
The fluorescence pattern induced by haematoxylin-eosin (HE) and Masson's trichrome (MT) staining methods on paraffin sections of rat embryos (from 13 to 18 days old) has been studied. Using optimal excitation (green light, 545 nm), HE- or MT-stained sections showed a selective red emission of the acidophilic tissue components, which was due to eosin Y in the case of HE and to acid fuchsin and/or xylidine ponceau in the case of MT. The fluorescence intensity induced by these anionic dyes was variable and related to the substrate nature and the embryo age. Whereas in young embryos only the immature red blood cells showed a noticeable fluorescence, in the oldest embryos there were also other tissue components that selectively fluoresced with these dyes, in particular fibre lens cells, elastic fibres, zymogen granules and muscle cells. Spectrofluorometric studies on free dyes and densitometric analysis of protein blots confirmed microscopical observations. Our results indicate that the standard HE and MT staining methods can be used in recognizing the appearance of specific protein structures in embryonic tissues by means of fluorescence microscopy.
Sigma-Aldrich
Anti-β-Tubulin Antibody, clone AA2, Alexa Fluor 488 conjugate, clone AA2, Upstate®, from mouse