Merck
  • Cryptoxanthin structural isomers in oranges, orange juice, and other fruits.
Cryptoxanthin structural isomers in oranges, orange juice, and other fruits.
Journal of agricultural and food chemistry (2005-08-04)
Jörg Schlatterer, Dietmar E Breithaupt
ABSTRACT
Citrus fruits contain a wide range of bioactive compounds. Their carotenoid fraction is inter alia dominated by structural cryptoxanthin isomers as beta-cryptoxanthin and zeinoxanthin. Both xanthophylls were identified in saponified citrus fruit extracts by comparison to reference compounds extracted from corn and by their typical fragmentation pattern in LC-(APCI)MS analyses. alpha-Cryptoxanthin, another structural cryptoxanthin isomer usually found in carrot leaves, was not identified in the citrus fruits studied. Cryptoxanthin concentrations of direct orange juices (D) and reconstituted juices (C) were compared. Although the respective mean values [beta-cryptoxanthin, 62 (C) versus 110 microg/100 g (D); zeinoxanthin, 22 (C) versus 37 microg/100 g (D)] were statistically distinguishable (P < 0.05%), a doubtless classification is not possible because the concentration ranges overlap. To identify esters of structural cryptoxanthin isomers in native orange juice extracts, four saturated acyl esters were synthesized. LC-(APCI)MS studies revealed for the first time that the dominant acylation partners of both xanthophylls were C12:0, C14:0, and C16:0 in nearly equal amounts of roughly one-third, whereas C10:0 and C18:1 were present at lower extents of 5-14%; other acylation partners were not identified. The presented method is appropriate to gain deeper insight into the pattern of structural cryptoxanthin isomers of citrus fruits. Knowledge of acylated cryptoxanthin isomers may be important in the evaluation of the bioavailability of individual esters in future human digestion studies.
Sigma-Aldrich
α-Cryptoxanthin, ≥90.0% (HPLC)