Merck
  • A single amino acid residue controls acyltransferase activity in a polyketide synthase from Toxoplasma gondii.
A single amino acid residue controls acyltransferase activity in a polyketide synthase from Toxoplasma gondii.
iScience (2022-07-26)
Hannah K D'Ambrosio, Jack G Ganley, Aaron M Keeler, Emily R Derbyshire
ABSTRACT
Type I polyketide synthases (PKSs) are multidomain, multimodule enzymes capable of producing complex polyketide metabolites. These modules contain an acyltransferase (AT) domain, which selects acyl-CoA substrates to be incorporated into the metabolite scaffold. Herein, we reveal the sequences of three AT domains from a polyketide synthase (TgPKS2) from the apicomplexan parasite Toxoplasma gondii. Phylogenic analysis indicates these ATs (AT1, AT2, and AT3) are distinct from domains in well-characterized microbial biosynthetic gene clusters. Biochemical investigations revealed that AT1 and AT2 hydrolyze malonyl-CoA but the terminal AT3 domain is non-functional. We further identify an "on-off switch" residue that controls activity such that a single amino acid change in AT3 confers hydrolysis activity while the analogous mutation in AT2 eliminates activity. This biochemical analysis of AT domains from an apicomplexan PKS lays the foundation for further molecular and structural studies on PKSs from T. gondii and other protists.
Sigma-Aldrich
Methylmalonyl coenzyme A tetralithium salt hydrate, ≥90% (HPLC)
Sigma-Aldrich
Malonyl coenzyme A lithium salt, ≥90% (HPLC)