Merck
  • Photoresponsive gels prepared by ring-opening metathesis polymerization.
Photoresponsive gels prepared by ring-opening metathesis polymerization.
Macromolecular rapid communications (2013-11-12)
Patricia Gumbley, Xiaoran Hu, John A Lawrence, Samuel W Thomas
ABSTRACT
This communication describes photoresponsive gels, prepared using ring-opening metathesis polymerization (ROMP), that dissolve upon irradiation with ultraviolet light. Exposure of mixtures of norbornene-type ROMP monomers and new photoreactive cross-linkers comprising two norbornene units bound through a chain containing o-nitrobenzyl esters (NBEs) to well-known ruthenium carbene catalysts gave cross-linked polymer networks that swelled in organic solvents or water depending on the structure of the monomer. These gels became homogeneous upon irradiation with UV light, consistent with breaking of the cross-links through photolysis of the NBE groups. The irradiation time required for homogenization of the gels depended on the cross-link density and the structure of the photoresponsive cross-linker.
Sigma-Aldrich
exo-5-Norbornenecarboxylic acid, 97%