Merck
  • Real-Time In Vivo Hepatotoxicity Monitoring through Chromophore-Conjugated Photon-Upconverting Nanoprobes.
Real-Time In Vivo Hepatotoxicity Monitoring through Chromophore-Conjugated Photon-Upconverting Nanoprobes.
Angewandte Chemie (International ed. in English) (2017-03-16)
Juanjuan Peng, Animesh Samanta, Xiao Zeng, Sanyang Han, Lu Wang, Dongdong Su, Daniel Teh Boon Loong, Nam-Young Kang, Sung-Jin Park, Angelo Homayoun All, Wenxuan Jiang, Lin Yuan, Xiaogang Liu, Young-Tae Chang
ABSTRACT
Drug toxicity is a long-standing concern of modern medicine. A typical anti-pain/fever drug paracetamol often causes hepatotoxicity due to peroxynitrite ONOO- . Conventional blood tests fail to offer real-time unambiguous visualization of such hepatotoxicity in vivo. Here we report a luminescent approach to evaluate acute hepatotoxicity in vivo by chromophore-conjugated upconversion nanoparticles. Upon injection, these nanoprobes mainly accumulate in the liver and the luminescence of nanoparticles remains suppressed owing to energy transfer to the chromophore. ONOO- can readily bleach the chromophore and thus recover the luminescence, the presence of ONOO- in the liver leads to fast restoring of the near-infrared emission. Taking advantages of the high tissue-penetration capability of near-infrared excitation/emission, these nanoprobes achieve real-time monitoring of hepatotoxicity in living animals, thereby providing a convenient screening strategy for assessing hepatotoxicity of synthetic drugs.
Sigma-Aldrich
Yttrium(III) acetate tetrahydrate, 99.99% trace rare earth metals basis