Merck
A catalytic asymmetric route to carbapenems.
Organic letters (2009-07-21)
Micah J Bodner, Ryan M Phelan, Craig A Townsend
ABSTRACT
Efficient syntheses of N-acetyl thienamycin and epithienamycin A in their readily deprotected form are reported where three contiguous stereocenters are established in a single catalytic asymmetric azetidinone-forming reaction. These examples are a template for synthesizing C-5/C-6 cis or trans carbapenems with independent control of the C-8 stereocenter. A library of oxidatively and sterochemically defined azetidinone precursors to a variety of naturally occurring carbapenems and potential biosynthetic intermediates has been prepared to facilitate studies of carbapenem antibiotic biosynthesis.
Sigma-Aldrich
N-Acetylcysteamine, 95%