Merck
  • Ozone Exerts Cytoprotective and Anti-Inflammatory Effects in Cardiomyocytes and Skin Fibroblasts after Incubation with Doxorubicin.
Ozone Exerts Cytoprotective and Anti-Inflammatory Effects in Cardiomyocytes and Skin Fibroblasts after Incubation with Doxorubicin.
Evidence-based complementary and alternative medicine : eCAM (2019-12-13)
V Simonetti, V Quagliariello, M Franzini, R V Iaffaioli, N Maurea, L Valdenassi
ABSTRACT
Skin reactions and cardiotoxicity are one of the most common side effects of doxorubicin in cancer patients. The main mechanisms based on the etiopathogenesis of these reactions are mediated by the overproduction of proinflammatory cytokines, metalloproteases, and the disruption of mitochondrial homeostasis. Ozone therapy demonstrated anti-inflammatory effects in several preclinical and clinical studies. The aim of this research is based on the evaluation of cardioprotective and dermatoprotective effects of ozone during incubation with doxorubicin, giving preliminary evidences for further studies in the field of cardio-oncology. Human skin fibroblast cells and human fetal cardiomyocytes were exposed to doxorubicin at subclinical concentration (100 nM) alone or combined with ozone concentrated from 10 up to 50 μg/mL. Cell viability and multiple anti-inflammatory studies were performed in both cell lines, with particular attention on the quantification of interleukins, leukotriene B4, NF-κB, and Nrf2 expressions during treatments. Ozone decreased significantly the cytotoxicity of doxorubicin in skin fibroblasts and cardiomyocytes after 24 h of incubation. The best cytoprotective effect of ozone was reached to 30 μg/mL with a plateau phase at higher concentration. Ozone also demonstrated anti-inflammatory effects decreasing significantly the interleukins and proinflammatory mediators in both cells. Ozone exerts cardioprotective and dermatoprotective effects during incubation with doxorubicin, and the involved mechanisms are mediated by its anti-inflammatory effects. The overall picture described herein is a pilot study for preclinical studies in oncology.
Sigma-Aldrich
HL-1 Cardiac Muscle Cell Line, HL-1 Cardiac Muscle Cell Line has been extensively characterized and is a valuable model system to address questions of cardiac biology at the cellular & molecular levels.