Merck
  • Control of Foodborne Staphylococcus aureus by Shikonin, a Natural Extract.
Control of Foodborne Staphylococcus aureus by Shikonin, a Natural Extract.
Foods (Basel, Switzerland) (2021-12-25)
Yangli Wan, Xiaowen Wang, Pengfei Zhang, Meng Zhang, Mingying Kou, Chao Shi, Xiaoli Peng, Xin Wang
ABSTRACT
Foodborne Staphylococcus aureus (S. aureus) has attracted widespread attention due to its foodborne infection and food poisoning in human. Shikonin exhibits antibacterial activity against a variety of microorganisms, but there are few studies on its antibacterial activity against S. aureus. This study aims to explore the antibacterial activity and mechanism of shikonin against foodborne S. aureus. The results show that the minimum inhibitory concentrations (MICs) and the minimum bactericidal concentrations (MBCs) of shikonin were equal for all tested strains ranging from 35 μg/mL to 70 μg/mL. Shikonin inhibited the growth of S. aureus by reducing intracellular ATP concentrations, hyperpolarizing cell membrane, destroying the integrity of cell membrane, and changing cell morphology. At the non-inhibitory concentrations (NICs), shikonin significantly inhibited biofilm formation of S. aureus, which was attributed to inhibiting the expression of cidA and sarA genes. Moreover, shikonin also markedly inhibited the transcription and expression of virulence genes (sea and hla) in S. aureus. In addition, shikonin has exhibited antibacterial ability against both planktonic and biofilm forms of S. aureus. Importantly, in vivo results show that shikonin has excellent biocompatibility. Moreover, both the heat stability of shikonin and the antimicrobial activity of shikonin against S. aureus were excellent in food. Our findings suggest that shikonin are promising for use as a natural food additive, and it also has great potential in effectively controlling the contamination of S. aureus in food and reducing the number of illnesses associated with S. aureus.
Sigma-Aldrich
Anti-Staphylococcal Enterotoxin A antibody produced in rabbit, whole antiserum
Pricing and availability is not currently available.