Merck
  • A Comprehensive Investigation of Steroidogenic Signaling in Classical and New Experimental Cell Models of Adrenocortical Carcinoma.
A Comprehensive Investigation of Steroidogenic Signaling in Classical and New Experimental Cell Models of Adrenocortical Carcinoma.
Cells (2022-05-15)
Sandra Sigala, Christina Bothou, David Penton, Andrea Abate, Mirko Peitzsch, Deborah Cosentini, Guido A M Tiberio, Stefan R Bornstein, Alfredo Berruti, Constanze Hantel
ABSTRACT
Adrenocortical carcinoma is a heterogeneous and aggressive cancer that originates from steroidogenic cells within the adrenal cortex. In this study, we have assessed for the preclinical gold standard NCI-H295 in direct comparison with the more recently established MUC-1 and a here newly reported ACC cell line (TVBF-7) the mutational status of important driver genes (TP53, MEN1, PRKAR1A, CTNNB1, APC, ZNRF-3, IGF-2, EGFR, RB1, BRCA1, BRCA2, RET, GNAS and PTEN), Wnt-signaling specificities (CTNNB1 mutation vs. APC mutation vs. wildtype), steroidogenic-(CYP11A1, CYP17A1, HSD3B2, HSD17B4, CYP21A2, CYP11B1, CYP11B2, MC2R, AT1R) and nuclear-receptor-signaling (AR, ER, GCR), varying electrophysiological potentials as well as highly individual hormone secretion profiles (Cortisol, Aldosterone, DHEA, DHEAS, Testosterone, 17-OH Progesterone, among others) which were investigated under basal and stimulated conditions (ACTH, AngII, FSK). Our findings reveal important genetic and pathophysiological characteristics for these three cell lines and reveal the importance of such cell-line panels reflecting differential endocrine functionalities to thereby better reflect clinically well-known ACC patient heterogeneities in preclinical studies.
Sigma-Aldrich
Phosphatase Inhibitor Cocktail 2, aqueous solution (dark coloration may develop upon storage, which does not affect the activity)
Sigma-Aldrich
Adrenocorticotropic Hormone human, ≥97% (HPLC)
Roche
cOmplete, Mini, EDTA-free Protease Inhibitor Cocktail, Protease Inhibitor Cocktail Tablets provided in a glass vial, Tablets provided in a glass vial