Merck
Haematology and blood chemistry values for several flamingo species.
Avian pathology : journal of the W.V.P.A (1992-01-01)
V I Peinado, F J Polo, G Viscor, J Palomeque
ABSTRACT
Reference values for some haematological and plasma chemical values in four species of clinically normal adult flamingos were established for use in avian medicine. The following variables were studied in rosy, greater, Chilean and lesser flamingos: haematocrit, haemoglobin concentration, erythrocyte and leucocyte counts, haematimetric indices, erythrocyte dimensions, glucose, urea, uric acid, cholesterol, creatinine, total bilirubin, aspartate aminotransferase, alanine aminotransferase, alkaline phosphatase, creatinine, phosphokinase, lactic dehydrogenase, total phosphorus, chloride, total plasma protein, albumin, globulins, albumin-globulin ratio, sodium, potassium, calcium, magnesium and osmolality.
Sigma-Aldrich
JNK1α1/SAPK1c Protein, inactive, 50 g, Unactive, N-terminal His6-tagged recombinant, human, full-length JNK1α1/SAPK1c, for use in Kinase Assays.