Merck
  • RIP3 antagonizes a TSC2-mediated pro-survival pathway in glioblastoma cell death.
RIP3 antagonizes a TSC2-mediated pro-survival pathway in glioblastoma cell death.
Biochimica et biophysica acta. Molecular cell research (2016-12-17)
Gregory Fettweis, Emmanuel Di Valentin, Laurent L'homme, Cédric Lassence, Franck Dequiedt, Marianne Fillet, Isabelle Coupienne, Jacques Piette
ABSTRACT
Glioblastomas are the deadliest type of brain cancer and are frequently associated with poor prognosis and a high degree of recurrence despite removal by surgical resection and treatment by chemo- and radio-therapy. Photodynamic therapy (PDT) is a treatment well known to induce mainly necrotic and apoptotic cell death in solid tumors. 5-Aminolevulinic acid (5-ALA)-based PDT was recently shown to sensitize human glioblastoma cells (LN-18) to a RIP3 (Receptor Interacting Protein 3)-dependent cell death which is counter-acted by activation of autophagy. These promising results led us to investigate the pathways involved in cell death and survival mechanisms occurring in glioblastoma following PDT. In the present study, we describe a new TSC2 (Tuberous Sclerosis 2)-dependent survival pathway implicating MK2 (MAPKAPK2) kinase and 14-3-3 proteins which conducts to the activation of a pro-survival autophagy. Moreover, we characterized a new RIP3/TSC2 complex where RIP3 is suggested to promote cell death by targeting TSC2-dependent survival pathway. These results highlight (i) a new role of TSC2 to protect glioblastoma against PDT-induced cell death and (ii) TSC2 and 14-3-3 as new RIP3 partners.
Sigma-Aldrich
SB 203580, solid, ≥98% (HPLC)
Sigma-Aldrich
MK-2 Inhibitor III, The MK-2 Inhibitor III, also referenced under CAS 1186648-22-5, controls the biological activity of MK-2. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.