Merck
  • Rab33b-exocyst interaction mediates localized secretion for focal adhesion turnover and cell migration.
Rab33b-exocyst interaction mediates localized secretion for focal adhesion turnover and cell migration.
iScience (2022-05-07)
Synne Arstad Bjørnestad, Noemi Antonella Guadagno, Ingrid Kjos, Cinzia Progida
ABSTRACT
Rab proteins are well known regulators of intracellular trafficking; however, more and more studies point to their function also in other cellular processes, including cell migration. In this work, we have performed an siRNA screen to identify Rab proteins that influence cell migration. The screen revealed Rab33b as the strongest candidate that affected cell motility. Rab33b has been previously reported to localize at the Golgi apparatus to regulate Golgi-to-ER retrograde trafficking and Golgi homeostasis. We revealed that Rab33b also mediates post-Golgi transport to the plasma membrane. We further identified Exoc6, a subunit of the exocyst complex, as an interactor of Rab33b. Moreover, our data indicate that Rab33b regulates focal adhesion dynamics by modulating the delivery of cargo such as integrins to focal adhesions. Altogether, our results demonstrate a role for Rab33b in cell migration by regulating the delivery of integrins to focal adhesions through the interaction with Exoc6.
Sigma-Aldrich
Monoclonal Anti-Vinculin antibody produced in mouse, clone hVIN-1, ascites fluid
Sigma-Aldrich
Anti-EXOC6 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Fibronectin human plasma, lyophilized powder, BioReagent, suitable for cell culture
Sigma-Aldrich
DAPI, for nucleic acid staining