Merck
  • Ubiquitin-dependent rapid degradation conceals a cell-protective function of cytoplasmic SIRT3 against oxidative stress.
Ubiquitin-dependent rapid degradation conceals a cell-protective function of cytoplasmic SIRT3 against oxidative stress.
Journal of biochemistry (2021-11-01)
Takashi Hayashi, Takashi Matsushita, Shin Hisahara, Naotoshi Iwahara, Atsushi Kuno, Risa Kunimoto, Ryusuke Hosoda, Masaya Tanno, Shun Shimohama, Yoshiyuki Horio
ABSTRACT
SIRT3 is an NAD+-dependent protein deacetylase localized in mitochondria. Several studies reported localization of SIRT3 in the cytoplasm or nucleus, but data of these studies were not consistent. We detected expression of mitochondrial (SIRT3mt) and cytoplasmic (SIRT3ct) Sirt3 mRNAs in the mouse brain, and we also found SIRT3 immunostaining of mitochondria and cytoplasm in the brain and cultured neural cells. However, expression levels of SIRT3ct in COS cells transfected with SIRT3ct cDNA were much lower than those of SIRT3mt. We found that SIRT3ct but not SIRT3mt was promptly degraded by ubiquitin-dependent degradation, in which SIRT3ct degradation was mediated mainly by ubiquitination of NH2-terminal methionine and partly by that of lysine residues of SIRT3ct. SIRT3ct expression level was significantly enhanced by the treatment of cells with staurosporine or H2O2. H2O2 treatment promoted nuclear translocation of SIRT3ct and induced histone H3 deacetylation and superoxide dismutase 2 expression. Overexpression of SIRT3ct decreased cell death caused by H2O2 at levels similar to those achieved by overexpression of SIRT3mt. Knockdown of Sirt3 mRNA increased cell death caused by amyloid-β (Aβ), and overexpression of SIRT3ct suppressed the toxic function of Aβ in PC12 cells. These results indicate that SIRT3ct promotes cell survival under physiological and pathological conditions.
Sigma-Aldrich
Anti-Histone H3, Acetylated (1-20) Rabbit pAb, liquid, Calbiochem®
Sigma-Aldrich
Anti-Beta III Tubulin Antibody, Chemicon®, from chicken
Sigma-Aldrich
Anti-GAPDH antibody, Mouse monoclonal, clone GAPDH-71.1, purified from hybridoma cell culture
Sigma-Aldrich
Monoclonal Anti-α-Tubulin antibody produced in mouse, ascites fluid, clone B-5-1-2
Sigma-Aldrich
Anti-Histone H3 antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
Millipore
ProteoExtract® Subcellular Proteome Extraction Kit