Merck
  • Antibody Conjugation of a Chimeric BET Degrader Enables in vivo Activity.
Antibody Conjugation of a Chimeric BET Degrader Enables in vivo Activity.
ChemMedChem (2019-11-02)
Thomas H Pillow, Pragya Adhikari, Robert A Blake, Jinhua Chen, Geoffrey Del Rosario, Gauri Deshmukh, Isabel Figueroa, Karen E Gascoigne, Amrita V Kamath, Susan Kaufman, Tracy Kleinheinz, Katherine R Kozak, Brandon Latifi, Douglas D Leipold, Chun Sing Li, Ruina Li, Melinda M Mulvihill, Aimee O'Donohue, Rebecca K Rowntree, Jack D Sadowsky, John Wai, Xinxin Wang, Cong Wu, Zijin Xu, Hui Yao, Shang-Fan Yu, Donglu Zhang, Richard Zang, Hongyan Zhang, Hao Zhou, Xiaoyu Zhu, Peter S Dragovich
ABSTRACT
The ability to selectively degrade proteins with bifunctional small molecules has the potential to fundamentally alter therapy in a variety of diseases. However, the relatively large size of these chimeric molecules often results in challenging physico-chemical properties (e. g., low aqueous solubility) and poor pharmacokinetics which may complicate their in vivo applications. We recently discovered an exquisitely potent chimeric BET degrader (GNE-987) which exhibited picomolar cell potencies but also demonstrated low in vivo exposures. In an effort to improve the pharmacokinetic properties of this molecule, we discovered the first degrader-antibody conjugate by attaching GNE-987 to an anti-CLL1 antibody via a novel linker. A single IV dose of the conjugate afforded sustained in vivo exposures that resulted in antigen-specific tumor regressions. Enhancement of a chimeric protein degrader with poor in vivo properties through antibody conjugation thereby expands the utility of directed protein degradation as both a biological tool and a therapeutic possibility.
Sigma-Aldrich
(S,R,S)-AHPC-di-trimethylamide-dioxodisulfide-carbonate ester
Pricing and availability is not currently available.