Merck
  • Vibrio cholerae Infection Induces Strain-Specific Modulation of the Zebrafish Intestinal Microbiome.
Vibrio cholerae Infection Induces Strain-Specific Modulation of the Zebrafish Intestinal Microbiome.
Infection and immunity (2021-06-02)
Paul Breen, Andrew D Winters, Kevin R Theis, Jeffrey H Withey
ABSTRACT
Zebrafish (Danio rerio) is an attractive model organism to use for an array of scientific studies, including host-microbe interactions. Zebrafish contain a core (i.e., consistently detected) intestinal microbiome consisting primarily of Proteobacteria. Furthermore, this core intestinal microbiome is plastic and can be significantly altered due to external factors. Zebrafish are particularly useful for the study of aquatic microbes that can colonize vertebrate hosts, including Vibrio cholerae. As an intestinal pathogen, V. cholerae must colonize the intestine of an exposed host for pathogenicity to occur. Members of the resident intestinal microbial community likely must be reduced or eliminated by V. cholerae for colonization, and subsequent disease, to occur. Many studies have explored a variety of aspects of the pathogenic effects of V. cholerae on zebrafish and other model organisms but few have researched how a V. cholerae infection changes the resident intestinal microbiome. In this study, 16S rRNA gene sequencing was used to examine how five genetically diverse V. cholerae strains alter the intestinal microbiome following an infection. We found that V. cholerae colonization induced significant changes in the zebrafish intestinal microbiome. Notably, changes in the microbial profile were significantly different from each other, based on the particular strain of V. cholerae used to infect zebrafish hosts. We conclude that V. cholerae significantly modulates the zebrafish intestinal microbiota to enable colonization and that specific microbes that are targeted depend on the V. cholerae genotype.
Sigma-Aldrich
Schiff′s reagent for aldehydes, suitable for microscopy
Sigma-Aldrich
Mucin from porcine stomach, Type III, bound sialic acid 0.5-1.5 %, partially purified powder