Merck
  • Expression of estrogen receptor alpha is associated with pathogenesis and prognosis of human papillomavirus-positive oropharyngeal cancer.
Expression of estrogen receptor alpha is associated with pathogenesis and prognosis of human papillomavirus-positive oropharyngeal cancer.
International journal of cancer (2019-06-23)
Makoto Kano, Satoru Kondo, Naohiro Wakisaka, Kosho Wakae, Mituharu Aga, Makiko Moriyama-Kita, Kazuya Ishikawa, Takayoshi Ueno, Yosuke Nakanishi, Miyako Hatano, Kazuhira Endo, Hisashi Sugimoto, Kouichi Kitamura, Masamichi Muramatsu, Tomokazu Yoshizaki
ABSTRACT
Human papillomavirus (HPV) has been identified as a causative agent of cervical cancer and oropharyngeal cancer (OPC). Intriguingly, estrogen and HPV were shown to play synergistic roles in cervical carcinogenesis. We recently demonstrated that the apolipoprotein B mRNA-editing catalytic polypeptide 3 (APOBEC3, A3) family, which is inducible by estrogen, could lead to HPV DNA hypermutation and cause viral DNA integration. In the present study, we examined the relationships between estrogen-estrogen receptor α (ERα) and A3s in HPV-positive OPC. ERα expression was associated with HPV positivity in OPC biopsy samples using immunohistochemical analysis and reverse-transcription quantitative polymerase chain reaction. In addition, ERα was significantly associated with improved overall survival in HPV-positive OPC (hazard ratio, 0.26; p = 0.029). APOBEC3A (A3A) mRNA was induced by estrogen in HPV and ERα-positive OPC cells. Furthermore, A3A mRNA and protein expression were significantly higher in ERα-positive cases than in ERα-negative ones, among HPV-positive biopsy samples (p = 0.037 and 0.047). These findings suggest that A3A is associated with a good prognosis in ERα-positive OPC, and indicate the prognostic significance of ERα in HPV-positive OPC. This is the first study to demonstrate the prognostic role of ERα in HPV-positive OPC.