Merck
  • Chromosome intermingling-the physical basis of chromosome organization in differentiated cells.
Chromosome intermingling-the physical basis of chromosome organization in differentiated cells.
Nucleic acids research (2016-03-05)
Shovamayee Maharana, K Venkatesan Iyer, Nikhil Jain, Mallika Nagarajan, Yejun Wang, G V Shivashankar
ABSTRACT
Chromosome territories (CTs) in higher eukaryotes occupy tissue-specific non-random three-dimensional positions in the interphase nucleus. To understand the mechanisms underlying CT organization, we mapped CT position and transcriptional changes in undifferentiated embryonic stem (ES) cells, during early onset of mouse ES cell differentiation and in terminally differentiated NIH3T3 cells. We found chromosome intermingling volume to be a reliable CT surface property, which can be used to define CT organization. Our results show a correlation between the transcriptional activity of chromosomes and heterologous chromosome intermingling volumes during differentiation. Furthermore, these regions were enriched in active RNA polymerase and other histone modifications in the differentiated states. These findings suggest a correlation between the evolution of transcription program in modifying CT architecture in undifferentiated stem cells. This leads to the formation of functional CT surfaces, which then interact to define the three-dimensional CT organization during differentiation.
Sigma-Aldrich
Monoclonal Anti-BrdU antibody produced in mouse, clone BU-33, ascites fluid, Immunohistology Grade
Sigma-Aldrich
5-Bromouridine 5′-triphosphate sodium salt, ≥93%