Merck
Cu/N,N'-Dibenzyloxalamide-Catalyzed N-Arylation of Heteroanilines.
Organic letters (2019-08-16)
Zhixiang Chen, Dawei Ma
ABSTRACT
N,N'-Dibenzyloxalamide (DBO) was identified as a powerful ligand for promoting Cu-catalyzed coupling of heteroanilines with (hetero)aryl halides. For (hetero)aryl chlorides, the coupling reaction occurred at 130 °C with 5 mol % CuBr and 10 mol % DBO. For (hetero)aryl bromides/iodides, coupling reaction took place at 80-100 °C with 1 mol % CuI and 2 mol % DBO. A variety of heteroanilines worked well to afford the arylation products in good to excellent yields.
Sigma-Aldrich
DBO, ≥95%