Merck
Conglomerate crystallization on self-assembled monolayers.
Chemical communications (Cambridge, England) (2011-10-12)
Michal Ejgenberg, Yitzhak Mastai
ABSTRACT
In this communication, we demonstrate that chiral self-assembled monolayers can be used for polymorphism control of chiral crystals. We studied the crystallization of DL-glutamic acid on chiral self-assembled monolayers and showed that crystallization of DL-glutamic acid on the chiral SAMs resulted in stabilization of the metastable conglomerate form.
Sigma-Aldrich
L-Leucine methyl ester hydrochloride, 98%
Sigma-Aldrich
DL-Glutamic acid monohydrate, ≥98% (HPLC)