Merck
  • Tax-inducible production of CC chemokine ligand 22 by human T cell leukemia virus type 1 (HTLV-1)-infected T cells promotes preferential transmission of HTLV-1 to CCR4-expressing CD4+ T cells.
Tax-inducible production of CC chemokine ligand 22 by human T cell leukemia virus type 1 (HTLV-1)-infected T cells promotes preferential transmission of HTLV-1 to CCR4-expressing CD4+ T cells.
Journal of immunology (Baltimore, Md. : 1950) (2008-01-08)
Kunio Hieshima, Daisuke Nagakubo, Takashi Nakayama, Aiko-Konno Shirakawa, Zhe Jin, Osamu Yoshie
ABSTRACT
Adult T cell leukemia is a mature CD4+ T cell malignancy which predominantly expresses CCR4 and is etiologically associated with human T cell leukemia virus type 1 (HTLV-1). Because HTLV-1 transmission depends on close cell-cell contacts, HTLV-1-infected T cells may preferentially interact with CCR4+CD4+ T cells for efficient viral transmission. In terms of gene expression and protein secretion, we found a strong correlation between HTLV-1 Tax oncoprotein and CCL22, a CCR4 ligand, in HTLV-1-infected T cells. Transient Tax expression in an HTLV-1-negative T cell line activated the CCL22 promoter and induced CCL22. Additionally, tax gene knockdown by small interference RNA reduced CCL22 expression in the infected T cells. These findings indicate that CCL22 is a cellular target gene of Tax. In chemotaxis assays, the culture supernatants of HTLV-1-infected T cells selectively attracted CCR4+CD4+ T cells in PBMCs. This was blocked by pretreating the supernatants with anti-CCL22 Ab or PBMCs with a synthetic CCR4 antagonist. In coculture experiments, primary CCR4+CD4+ T cells significantly adhered to Tax-expressing cells. This adhesion was blocked by the CCR4 antagonist or pertussis toxin. Interestingly, CCR4 was redistributed to the contact region, and in some cases, this was accompanied by a polarized microtubule-organizing center, which is an indicator of virological synapse formation, in the infected T cells. Finally, anti-CCL22 Ab treatment also blocked HTLV-1 transmission to primary CD4+ T cells in coculture experiments with HTLV-1 producer cells. Thus, HTLV-1-infected T cells produce CCL22 through Tax and selectively interact with CCR4+CD4+ T cells, resulting in preferential transmission of HTLV-1 to CCR4+CD4+ T cells.
Sigma-Aldrich
Monoclonal Anti-Rabbit Immunoglobulins–FITC antibody produced in mouse, clone RG-16, purified immunoglobulin, buffered aqueous solution
Sigma-Aldrich
Anti-β-Tubulin−Cy3 antibody, Mouse monoclonal, clone TUB 2.1, purified from hybridoma cell culture