Merck
Quantification of Plant Cell Death by Electrolyte Leakage Assay.
Bio-protocol (2018-03-05)
Noriyuki Hatsugai, Fumiaki Katagiri
ABSTRACT
We describe a protocol to measure the electrolyte leakage from plant tissues, resulting from loss of cell membrane integrity, which is a common definition of cell death. This simple protocol is designed to measure the electrolyte leakage from a tissue sample over a time course, so that the extent of cell death in the tissue can be monitored dynamically. In addition, it is easy to handle many tissue samples in parallel, which allows a high level of biological replication. Although the protocol is exemplified by cell death in Arabidopsis in response to pathogen challenge, it is easily applicable to other types of plant cell death.
Sigma-Aldrich
Rifampicin, ≥95% (HPLC), powder or crystals