Merck
  • Virucidal and Neutralizing Activity Tests for Antiviral Substances and Antibodies.
Virucidal and Neutralizing Activity Tests for Antiviral Substances and Antibodies.
Bio-protocol (2018-05-20)
Chie Aoki-Utsubo, Ming Chen, Hak Hotta
ABSTRACT
In a narrow definition, virucidal activity represents the activity by which to interact with and physically disrupt viral particles. In a broad definition, it includes the activity by which to functionally inhibit (neutralize) viral infectivity without apparent morphological alterations of the viral particles. The viral infectivity can be measured in cell culture system by means of plaque assay, infectious focus assay, 50% tissue culture infectious dose (TCID50) assay, etc. Morphologically, disruption of viral particles can be demonstrated by negative staining electron microscopic analysis of viral particles. In this article, we describe methods to assess virucidal activity in a broad definition.