Merck
  • Dynamics of telomeric repeat-containing RNA expression in early embryonic cleavage stages with regards to maternal age.
Dynamics of telomeric repeat-containing RNA expression in early embryonic cleavage stages with regards to maternal age.
Aging (2020-08-30)
Paweł Kordowitzki, Isabel López de Silanes, Ana Guío-Carrión, Maria A Blasco
ABSTRACT
Telomeres are transcribed into long non-coding RNAs known as Telomeric Repeat-Containing RNA (TERRA). They have been shown to be essential regulators of telomeres and to act as epigenomic modulators at extra-telomeric sites. However the role of TERRA during early embryonic development has never been investigated. Here, we show that TERRA is expressed in murine and bovine early development following a wave pattern. It starts at 4-cell stage, reaching a maximum at the 16-cell followed by a decline at the morula and blastocyst stages. Moreover, TERRA expression is not affected by increasing oocyte donor age whereas telomere length does. This indicates that TERRA expression is independent of the telomere length in early development. Our findings anticipate an essential role of TERRA in early stages of development and this might be useful in the future for a better understanding of age related female infertility.
Sigma-Aldrich
Bovine Serum Albumin, heat shock fraction, pH 7, ≥98%
Sigma-Aldrich
Sodium pyruvate, powder, BioXtra, suitable for mouse embryo cell culture
Sigma-Aldrich
Epidermal Growth Factor from murine submaxillary gland, EGF, suitable for cell culture
Sigma-Aldrich
MEM Non-essential Amino Acid Solution (100×), without L-glutamine, liquid, sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
BME Amino Acids Solution 50×, Without L-glutamine, sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Phenol red solution, 0.5%, liquid, sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Medium 199, With Earle′s salts, L-glutamine and sodium bicarbonate, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Sodium DL-lactate solution, BioReagent, syrup, 60 % (w/w), synthetic, suitable for cell culture