Merck
  • RNA-binding protein SORBS2 suppresses clear cell renal cell carcinoma metastasis by enhancing MTUS1 mRNA stability.
RNA-binding protein SORBS2 suppresses clear cell renal cell carcinoma metastasis by enhancing MTUS1 mRNA stability.
Cell death & disease (2020-12-15)
Qi Lv, Fan Dong, Yong Zhou, Zhiping Cai, Gangmin Wang
ABSTRACT
RNA-binding proteins (RBPs) predominantly contribute to abnormal posttranscriptional gene modulation and disease progression in cancer. Sorbin and SH3 domain-containing 2 (SORBS2), an RBP, has been reported to be a potent tumor suppressor in several cancer types. Through integrative analysis of clinical specimens, we disclosed that the expression level of SORBS2 was saliently decreased in metastatic tissues and positively correlated with overall survival. We observed that overexpression of SORBS2 brought about decreased metastatic capacity in ccRCC cell lines. Transcriptome-wide analysis revealed that SORBS2 notably increased microtubule-associated tumor-suppressor 1 gene (MTUS1) expression. In-depth mechanistic exploring discovered that the Cys2-His2 zinc finger (C2H2-ZnF) domain of SORBS2 directly bound to the 3' untranslated region (3'UTR) of MTUS1 mRNA, which increased MTUS1 mRNA stability. In addition, we identified that MTUS1 regulated microtubule dynamics via promoting KIF2CS192 phosphorylation by Aurora B. Together, our research identified SORBS2 as a suppressor of ccRCC metastasis by enhancing MTUS1 mRNA stability, providing a novel understanding of RBPs during ccRCC progression.
Sigma-Aldrich
Monoclonal Anti-β-Tubulin antibody produced in mouse, clone TUB 2.1, ascites fluid
Sigma-Aldrich
Monoclonal Anti-Tubulin, Acetylated antibody produced in mouse, clone 6-11B-1, ascites fluid
Sigma-Aldrich
Anti-SORBS2 antibody,Mouse monoclonal, clone S5C, purified from hybridoma cell culture