Merck
  • Coordination of mitochondrial and cellular dynamics by the actin-based motor Myo19.
Coordination of mitochondrial and cellular dynamics by the actin-based motor Myo19.
Journal of cell science (2021-05-21)
Katarzyna Majstrowicz, Ulrike Honnert, Petra Nikolaus, Vera Schwarz, Stefanie J Oeding, Sandra A Hemkemeyer, Martin Bähler
ABSTRACT
Myosin XIX (Myo19) is an actin-based motor that competes with adaptors of microtubule-based motors for binding to the outer mitochondrial transmembrane proteins Miro1 and Miro2 (collectively Miro, also known as RhoT1 and RhoT2, respectively). Here, we investigate which mitochondrial and cellular processes depend on the coordination of Myo19 and microtubule-based motor activities. To this end, we created Myo19-deficient HEK293T cells. Mitochondria in these cells were not properly fragmented at mitosis and were partitioned asymmetrically to daughter cells. Respiratory functions of mitochondria were impaired and ROS generation was enhanced. On a cellular level, cell proliferation, cytokinesis and cell-matrix adhesion were negatively affected. On a molecular level, Myo19 regulates focal adhesions in interphase, and mitochondrial fusion and mitochondrially associated levels of fission protein Drp1 and adaptor proteins dynactin and TRAK1 at prometaphase. These alterations were due to a disturbed coordination of Myo19 and microtubule-based motor activities by Miro.
Sigma-Aldrich
(+)-S-Trityl-L-cysteine, 97%
Sigma-Aldrich
Monoclonal Anti-Phosphotyrosine antibody produced in mouse, clone PT-66, ascites fluid
Sigma-Aldrich
Monoclonal Anti-Vinculin antibody produced in mouse, clone hVIN-1, ascites fluid
Sigma-Aldrich
Anti-Dynein Antibody, 74 kDa Intermediate chains, cytoplasmic, clone 74.1, clone 74.1, Chemicon®, from mouse
Sigma-Aldrich
Poly-L-lysine hydrobromide, mol wt 70,000-150,000, lyophilized powder, γ-irradiated, BioXtra, suitable for cell culture
Sigma-Aldrich
Monoclonal Anti-β-Tubulin antibody produced in mouse, clone TUB 2.1, ascites fluid
Sigma-Aldrich
N-(2-Mercaptopropionyl)glycine
Sigma-Aldrich
Fibronectin human plasma, lyophilized powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Anti-β-Actin antibody, Mouse monoclonal, clone AC-15, purified from hybridoma cell culture