Merck
  • IL-1β/TNF-α/IL-6 inflammatory cytokines promote STAT1-dependent induction of CH25H in Zika virus-infected human macrophages.
IL-1β/TNF-α/IL-6 inflammatory cytokines promote STAT1-dependent induction of CH25H in Zika virus-infected human macrophages.
The Journal of biological chemistry (2019-08-04)
Tshifhiwa Magoro, Aditya Dandekar, Lucas T Jennelle, Rohan Bajaj, Gabriel Lipkowitz, Angelina R Angelucci, Pascal O Bessong, Young S Hahn
ABSTRACT
Zika virus (ZIKV)3 is an enveloped, single-stranded, positive-sense RNA virus of the Flaviviridae family that has emerged as a public health threat because of its global transmission and link to microcephaly. Currently there is no vaccine for this virus. Conversion of cholesterol to 25-hydroxycholesterol by cholesterol 25-hydroxylase (CH25H) has been shown to have broad antiviral properties. However, the molecular basis of induction of CH25H in humans is not known. Elucidation of signaling and transcriptional events for induction of CH25H expression is critical for designing therapeutic antiviral agents. In this study, we show that CH25H is induced by ZIKV infection or Toll-like receptor stimulation. Interestingly, CH25H is induced by pro-inflammatory cytokines, including IL-1β, tumor necrosis factor α, and IL-6, and this induction depends on the STAT1 transcription factor. Additionally, we observed that cAMP-dependent transcription factor (ATF3) weakly binds to the CH25H promoter, suggesting cooperation with STAT1. However, ZIKV-induced CH25H was independent of type I interferon. These findings provide important information for understanding how the Zika virus induces innate inflammatory responses and promotes the expression of anti-viral CH25H protein.
Sigma-Aldrich
TLR3/dsRNA Complex Inhibitor, The TLR3/dsRNA Complex Inhibitor blocks dsRNA binding to TLR3.