Merck
  • Flow cytometry multiplexed method for the detection of neutralizing human antibodies to the native SARS-CoV-2 spike protein.
Flow cytometry multiplexed method for the detection of neutralizing human antibodies to the native SARS-CoV-2 spike protein.
EMBO molecular medicine (2021-01-21)
Lydia Horndler, Pilar Delgado, David Abia, Ivaylo Balabanov, Pedro Martínez-Fleta, Georgina Cornish, Miguel A Llamas, Sergio Serrano-Villar, Francisco Sánchez-Madrid, Manuel Fresno, Hisse M van Santen, Balbino Alarcón
ABSTRACT
A correct identification of seropositive individuals for the severe acute respiratory syndrome coronavirus-2 (SARS-CoV-2) infection is of paramount relevance to assess the degree of protection of a human population to present and future outbreaks of the COVID-19 pandemic. We describe here a sensitive and quantitative flow cytometry method using the cytometer-friendly non-adherent Jurkat T-cell line that stably expresses the full-length native spike "S" protein of SARS-CoV-2 and a truncated form of the human EGFR that serves a normalizing role. S protein and huEGFRt coding sequences are separated by a T2A self-cleaving sequence, allowing to accurately quantify the presence of anti-S immunoglobulins by calculating a score based on the ratio of fluorescence intensities obtained by double-staining with the test sera and anti-EGFR. The method allows to detect immune individuals regardless of the result of other serological tests or even repeated PCR monitoring. As examples of its use, we show that as much as 28% of the personnel working at the CBMSO in Madrid is already immune. Additionally, we show that anti-S antibodies with protective neutralizing activity are long-lasting and can be detected in sera 8 months after infection.
Sigma-Aldrich
Anti-2A peptide Antibody, clone 3H4, clone 3H4, from mouse