Merck
  • Aldehyde oxidase assay by capillary electrophoresis: From off-line, online up to immobilized enzyme reactor.
Aldehyde oxidase assay by capillary electrophoresis: From off-line, online up to immobilized enzyme reactor.
Journal of separation science (2020-07-07)
Shengyun Huang, Andrea Celá, Erwin Adams, Zdenĕk Glatz, Ann Van Schepdael
ABSTRACT
Capillary electrophoresis is a modern separation technique characterized by many benefits, which qualify it also for enzyme assays and the study of enzyme kinetics during drug development. Homogeneous or heterogeneous approaches can be followed for the enzymatic incubation. In this study, an immobilization procedure of aldehyde oxidase on magnetic particles was developed considering their integration with capillary electrophoresis. A number of magnetic nano/microparticle types were tested for this purpose, showing that aldehyde oxidase was most active when immobilized on bare silica magnetic nanoparticles. Primarily, the reusability of the enzyme immobilized on bare silica nanoparticles was tested. Three consecutive incubations with substrate could be performed, but the activity considerably dropped after the first incubation. One reason could be an enzyme detachment from the nanoparticles, but no release was detected neither at 4°C nor at 37°C during 5 h. The drop in enzymatic activity observed in consecutive incubations, could also be due to inactivation of the enzyme over time at given temperature. For the immobilized enzyme stored at 4°C, the activity decreased to 83% after 5 h, in contrast with a steep decrease at 37°C to 37%.
Sigma-Aldrich
Phthalazone, 99%
Sigma-Aldrich
N-(3-Dimethylaminopropyl)-N′-ethylcarbodiimide, ≥97.0% (T)