Merck
  • Fine particulate matter inhibits phagocytosis of macrophages by disturbing autophagy.
Fine particulate matter inhibits phagocytosis of macrophages by disturbing autophagy.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology (2020-10-31)
Ying Li, Yu-Le Yong, Menglin Yang, Weijia Wang, Xiaoyan Qu, Xiaomin Dang, Dong Shang, Yongping Shao, Jiankang Liu, Ying Chang
ABSTRACT
Mounting evidence from epidemiological and clinical studies has revealed marked correlations between the air pollutant fine particulate matter (FPM) and respiratory diseases. FPM reaches distal airways and deposits in alveolar regions where it can act directly on alveolar macrophages. However, the detailed effect of FPM on the physiological function of alveolar macrophages and the underlying mechanisms remain unclear. In this study, we showed that exposing THP-1-derived macrophages to FPM led to autophagy dysfunction. FPM activated the extracellular signal-regulated kinase 1/2 (ERK1/2) signaling pathway, which promoted the expression of autophagy-related 2A (ATG2A) and reactive oxygen species generation. The overexpression of ATG2A enhanced the synthesis of autophagic membranes, and the excessive production of reactive oxygen species caused autophagy flux inhibition through disrupting the lysosomal activity. More importantly, FPM impaired the phagocytic ability of macrophages on Escherichia coli and apoptotic neutrophils. Finally, we showed that restoring autophagy rescued the impairment of phagocytic ability induced by FPM. In summary, these results reveal the molecular mechanism of autophagy dysfunction caused by FPM and provide a novel approach to resolve the impaired function of macrophages in respiratory diseases induced by FPM.
Sigma-Aldrich
SP600125, ≥98% (HPLC)
Sigma-Aldrich
3-Methyladenine, autophagy inhibitor
Sigma-Aldrich
Acridine Orange hydrochloride solution, 10 mg/mL in H2O, ≥95.0% (HPLC)
Sigma-Aldrich
2′,7′-Dichlorodihydrofluorescein diacetate, ≥97%
Sigma-Aldrich
MISSION® esiRNA, targeting human ATG2A