Merck
  • The Effect of Korean Red Ginseng on Bisphenol A-Induced Fatty Acid Composition and Lipid Metabolism-Related Gene Expression Changes.
The Effect of Korean Red Ginseng on Bisphenol A-Induced Fatty Acid Composition and Lipid Metabolism-Related Gene Expression Changes.
The American journal of Chinese medicine (2020-12-11)
Joonwoo Park, KeunOh Choi, Jeonggeun Lee, Jong-Min Jung, YoungJoo Lee
ABSTRACT
Bisphenol A (BPA), which is known to be an endocrine-disrupting chemical (EDC), is associated not only with estrogen activity and reproductive toxicity but also with a variety of metabolic disorders. BPA affects glucose tolerance, cholesterol biosynthesis, and fatty acid synthesis. Ginseng is a traditional medicinal plant that has been widely used in East Asia for more than 2000 years, and a number of health effects have been reported. Korean Red Ginseng (KRG) has also been shown to have effects on lipid metabolism and body weight reduction in vivo in obese mice. In this study, we administered BPA and KRG to ovariectomized (OVX) ICR mice. BPA (800 mg/kg/day) and KRG (1.2 g/kg/day) were orally administered to OVX mice for 3 days. KRG inhibited the increase in total fatty acid level by BPA as determined by lipid profiling in the liver of OVX mice. In addition, transcriptome analysis showed that KRG inhibited BPA-induced changes in lipid metabolic process-related genes. Our findings suggest that KRG can regulate BPA-induced changes in lipid metabolism.
Product Description
Supelco
Silica gel, high-purity grade (Davisil Grade 635), pore size 60 Å, 60-100 mesh