Merck
  • Anti-biofilm Fe3O4@C18-[1,3,4]thiadiazolo[3,2-a]pyrimidin-4-ium-2-thiolate Derivative Core-shell Nanocoatings.
Anti-biofilm Fe3O4@C18-[1,3,4]thiadiazolo[3,2-a]pyrimidin-4-ium-2-thiolate Derivative Core-shell Nanocoatings.
Materials (Basel, Switzerland) (2020-10-22)
Rodica Olar, Mihaela Badea, Cătălin Maxim, Alexandru Mihai Grumezescu, Coralia Bleotu, Luminiţa Măruţescu, Mariana Carmen Chifiriuc
ABSTRACT
The derivatives 5,7-dimethyl[1,3,4]thiadiazolo[3,2-a]pyrimidin-4-ium-2-thiolate (1) and 7-methyl-5-phenyl[1,3,4]thiadiazolo[3,2-a]pyrimidin-4-ium-2-thiolate (2) were fully characterized by single-crystal X-ray diffraction. Their supramolecular structure is built through both π-π stacking and C=S-π interactions for both compounds. The embedment of the tested compounds into Fe3O4@C18 core-shell nanocoatings increased the protection degree against Candida albicans biofilms on the catheter surface, suggesting that these bioactive nanocoatings could be further developed as non-cytotoxic strategies for fighting biofilm-associated fungal infections.
Sigma-Aldrich
5-Amino-1,3,4-thiadiazole-2-thiol, 95%