Merck
  • Endocrine-Disrupting Organochlorine Pesticides in Human Breast Milk: Changes during Lactation.
Endocrine-Disrupting Organochlorine Pesticides in Human Breast Milk: Changes during Lactation.
Nutrients (2021-01-21)
Agata Witczak, Anna Pohoryło, Hassan Abdel-Gawad
ABSTRACT
The aim of the present study was to assess infant safety associated with the occurrence of endocrine-disrupting organochlorine pesticides (OCP) in breast milk. Moreover, the association between pregnant mothers' dietary habits and these compounds levels in breast milk was investigated. Breast milk was collected at various stages of lactation. The samples were analyzed by the GC-MS method. The OCP concentrations ranged from < limit of detection (LOD) to 6.81 ng/g lipids. The highest OCP concentrations in breast milk occurred primarily within the first month of lactation, and decreased over the lactation period. It was found that the maternal consumption of certain food products-in particular pork, beef, poultry, eggs, and dairy products-could have affected the content of 1,1'-(2,2,2-Trichloroethane-1,1-diyl)bis(4-chlorobenzene), called DDT and its metabolites in the breast milk. The levels of beta-endosulfan were positively correlated with fish and poultry consumption. The redundancy analysis indicated that the diets of the pregnant women had an important impact on pesticide residues in the breast milk. There is a potential possibility of lowering the content of organochlorine compounds in breast milk by adhering to nutritional recommendations, e.g., avoiding the excessive consumption of fish and other raw food materials of unknown origin.
Product Description
Supelco
Endosulfan, PESTANAL®, analytical standard
Pricing and availability is not currently available.
Supelco
HCH, PESTANAL®, analytical standard, mixture of isomers (α:β:γ:δ=1:1:1:1)
Pricing and availability is not currently available.