Merck
  • Elimination of GlnKAmtB affects serine biosynthesis and improves growth and stress tolerance of Escherichia coli under nutrient-rich conditions.
Elimination of GlnKAmtB affects serine biosynthesis and improves growth and stress tolerance of Escherichia coli under nutrient-rich conditions.
FEMS microbiology letters (2020-11-27)
Romina Frare, Margarita Stritzler, Cecilia Pascuan, Karen Liebrenz, Luisa Galindo-Sotomonte, Gabriela Soto, Pablo Iván Nikel, Nicolás Ayub
ABSTRACT
Nitrogen is a most important nutrient resource for Escherichia coli and other bacteria that harbor the glnKamtB operon, a high-affinity ammonium uptake system highly interconnected with cellular metabolism. Although this system confers an advantage to bacteria when growing under nitrogen-limiting conditions, little is known about the impact of these genes on microbial fitness under nutrient-rich conditions. Here, the genetically tractable E. coli BW25113 strain and its glnKamtB-null mutant (JW0441) were used to analyze the impact of GlnK-AmtB on growth rates and oxidative stress tolerance. Strain JW0441 showed a shorter initial lag phase, higher growth rate, higher citrate synthase activity, higher oxidative stress tolerance and lower expression of serA than strain BW25113 under nutrient-rich conditions, suggesting a fitness cost to increase metabolic plasticity associated with serine metabolism. The overexpression of serA in strain JW0441 resulted in a decreased growth rate and stress tolerance in nutrient-rich conditions similar to that of strain BW25113, suggesting that the negative influence on bacterial fitness imposed by GlnK-AmtB can be traced to the control of serine biosynthesis. Finally, we discuss the potential applications of glnKamtB mutants in bioproduction processes.
Sigma-Aldrich
Sodium phosphate dibasic, 99.95% trace metals basis
Sigma-Aldrich
Potassium phosphate monobasic, for molecular biology, ≥98.0%
Sigma-Aldrich
Citrate Synthase Assay Kit, 1 kit sufficient for 100 reactions (using a 1 ml cuvette), 1 kit sufficient for 480 reactions (using 96 multiwell plates)
Sigma-Aldrich
Ammonium chloride, ACS reagent, ≥99.5%