Merck
  • Melatonin sensitises shikonin-induced cancer cell death mediated by oxidative stress via inhibition of the SIRT3/SOD2-AKT pathway.
Melatonin sensitises shikonin-induced cancer cell death mediated by oxidative stress via inhibition of the SIRT3/SOD2-AKT pathway.
Redox biology (2020-08-31)
Mengling Li, Chengai Wu, Jibran Sualeh Muhammad, Dan Yan, Koichi Tsuneyama, Hideki Hatta, Zheng-Guo Cui, Hidekuni Inadera
ABSTRACT
Recent research suggests that melatonin (Mel), an endogenous hormone and natural supplement, possesses anti-proliferative effects and can sensitise cells to anti-cancer therapies. Although shikonin (SHK) also possesses potential anti-cancer properties, the poor solubility and severe systemic toxicity of this compound hinders its clinical usage. In this study, we combined Mel and SHK, a potentially promising chemotherapeutic drug combination, with the aim of reducing the toxicity of SHK and enhancing the overall anti-cancer effects. We demonstrate for the first time that Mel potentiates the cytotoxic effects of SHK on cancer cells by inducing oxidative stress via inhibition of the SIRT3/SOD2-AKT pathway. Particularly, Mel-SHK treatment induced oxidative stress, increased mitochondrial calcium accumulation and reduced the mitochondrial membrane potential in various cancer cells, leading to apoptosis. This drug combination also promoted endoplasmic reticulum (ER) stress, leading to AKT dephosphorylation. In HeLa cells, Mel-SHK treatment reduced SIRT3/SOD2 expression and SOD2 activity, while SIRT3 overexpression dramatically reduced Mel-SHK-induced oxidative stress, ER stress, mitochondrial dysfunction and apoptosis. Hence, we propose the combination of Mel and SHK as a novel candidate chemotherapeutic regimen that targets the SIRT3/SOD2-AKT pathway in cancer.
Sigma-Aldrich
Tetramethylrhodamine methyl ester perchlorate, ≥95%
Pricing and availability is not currently available.
Sigma-Aldrich
4-Bromo-Resveratrol, ≥98% (HPLC)
Pricing and availability is not currently available.
Sigma-Aldrich
Amyloid Protein Non-Aβ Component, ≥80% (HPLC)
Pricing and availability is not currently available.
Sigma-Aldrich
4-Phenylbutyric acid, 99%
Pricing and availability is not currently available.