Merck
  • Effects of bovine lactoferrin and chitosan nanoparticles on serum biochemical indices, antioxidative enzymes, transcriptomic responses, and resistance of Nile tilapia against Aeromonas hydrophila.
Effects of bovine lactoferrin and chitosan nanoparticles on serum biochemical indices, antioxidative enzymes, transcriptomic responses, and resistance of Nile tilapia against Aeromonas hydrophila.
Fish & shellfish immunology (2021-02-09)
Mona M Abdel-Wahab, Nabil M Taha, Mohamed A Lebda, Mohamed S Elfeky, Hany M R Abdel-Latif
ABSTRACT
The present study was carried out to investigate the effects of dietary bovine lactoferrin (BLF) or chitosan nanoparticles (CHN) alone or in combinations on serum biochemical indices, antioxidative capacity, transcriptomic responses, non-specific immunity, and resistance of Nile tilapia (Oreochromis niloticus) against challenge with Aeromonas hydrophila. Fish were fed on the basal diet with no supplements and served as control (CTR), and six other experimental diets containing different levels of BLF (800 and 1200 mg per kg diet), CHN (500 and 1000 mg per kg diet), and their combinations (400 mg BLF plus 250 mg CHN per kg diet, and 600 mg BLF plus 500 mg CHN per kg diet) for 45 days. At the end of the experiment, serum, and tissue specimens (liver and kidney) were collected, fish in all groups were challenged with A. hydrophila and then observed for another ten days to calculate the RPS. Compared to the CTR group, no significant differences were recorded in TP, ALB, GLO, BUN, and CREAT values among all treatments. Serum LYZ, ALT, AST, and ALP enzyme activities were significantly increased in all experimental groups over the CTR (P < 0.05), and their highest values were recorded in the combined treatments. Moreover, dietary supplementation with CHN (1000 mg/kg) and combined treatments significantly increased the SOD, CAT, and GSH-Px enzyme activities compared to other groups (P < 0.05). The highest mRNA expression levels of IGF-1 gene in liver, and IL-1β, and IFN-γ genes in kidneys were found in CHN (1000 mg/kg) group and combined treatments more than other groups. Interestingly, no, or mild histopathological alterations were noticed in the hepatopancreas and posterior kidney of the treated groups. A significantly higher RPS was identified in the combined treatments challenged with A. hydrophila compared with the CTR group. This study exemplifies the positive impacts of dietary supplementation with BLF or CHN alone or combinations on the antioxidative status, immunity, and disease resistance of Nile tilapia.
Sigma-Aldrich
Lactoferrin from bovine milk, ≥85% (SDS-PAGE)