Merck
  • Circular RNA RSF1 promotes inflammatory and fibrotic phenotypes of irradiated hepatic stellate cell by modulating miR-146a-5p.
Circular RNA RSF1 promotes inflammatory and fibrotic phenotypes of irradiated hepatic stellate cell by modulating miR-146a-5p.
Journal of cellular physiology (2020-01-22)
Yuhan Chen, Baoying Yuan, Genwen Chen, Li Zhang, Yuan Zhuang, Hao Niu, Zhaochong Zeng
ABSTRACT
The role of circular RNA (circRNA) in radiation-induced liver disease (RILD) remains largely unknown. In this study, Ras-related C3 botulinum toxin substrate 1 (RAC1) was elevated in irradiated human hepatic stellate cell (HSC) line LX2, the important effector cell mediating RILD. Overexpression of RAC1 promotes cell proliferation, proinflammatory cytokines production, and α-smooth muscle actin expression, which were blocked by microRNA (miR)-146a-5p mimics. CircRNA RSF1 (circRSF1) was upregulated in irradiated LX2 cells and predicted to harbor binding site for miR-146a-5p. Biotinylated-RNA pull down and dual-luciferase reporter detection confirmed the direct interaction of circRSF1 and miR-146a-5p. Enforced expression of circRSF1 increased RAC1 expression by acting as miR-146a-5p sponge to inhibit miR-146a-5p activity, and thus enhanced the cell viability, and promoted inflammatory and fibrotic phenotype of irradiated LX2 cells. These findings indicate a functional regulatory axis composing of circRSF1, miR-146a-5p, and RAC1 in irradiated HSC, which may provide attractive therapeutic targets for RILD.
Sigma-Aldrich
MISSION® esiRNA, targeting human RSF1