Merck
  • FS222, a CD137/PD-L1 Tetravalent Bispecific Antibody, Exhibits Low Toxicity and Antitumor Activity in Colorectal Cancer Models.
FS222, a CD137/PD-L1 Tetravalent Bispecific Antibody, Exhibits Low Toxicity and Antitumor Activity in Colorectal Cancer Models.
Clinical cancer research : an official journal of the American Association for Cancer Research (2020-04-30)
Matthew A Lakins, Alexander Koers, Raffaella Giambalvo, Jose Munoz-Olaya, Robert Hughes, Emma Goodman, Sylwia Marshall, Francisca Wollerton, Sarah Batey, Daniel Gliddon, Mihriban Tuna, Neil Brewis
ABSTRACT
With the increased prevalence in checkpoint therapy resistance, there remains a significant unmet need for additional therapies for patients with relapsing or refractory cancer. We have developed FS222, a bispecific tetravalent antibody targeting CD137 and PD-L1, to induce T-cell activation to eradicate tumors without the current toxicity and efficacy limitations seen in the clinic. A bispecific antibody (FS222) was developed by engineering CD137 antigen-binding sites into the Fc region of a PD-L1 IgG1 mAb. T-cell activation by FS222 was investigated using multiple in vitro assays. The antitumor efficacy, survival benefit, pharmacodynamics, and liver pharmacology of a murine surrogate molecule were assessed in syngeneic mouse tumor models. Toxicology and the pharmacokinetic/pharmacodynamic profile of FS222 were investigated in a non-human primate dose-range finding study. We demonstrated simultaneous binding of CD137 and PD-L1 and showed potent T-cell activation across CD8+ T-cell activation assays in a PD-L1-dependent manner with a CD137/PD-L1 bispecific antibody, FS222. FS222 also activated T cells in a human primary mixed lymphocyte reaction assay, with greater potency than the monospecific mAb combination. FS222 showed no signs of liver toxicity up to 30 mg/kg in a non-human primate dose-range finding study. A surrogate molecule caused significant tumor growth inhibition and survival benefit, concomitant with CD8+ T-cell activation, in CT26 and MC38 syngeneic mouse tumor models. By targeting CD137 agonism to areas of PD-L1 expression, predominantly found in the tumor microenvironment, FS222 has the potential to leverage a focused, potent, and safe immune response augmenting the PD-(L)1 axis blockade.
Product Description
Ficoll® Paque Plus, Cytiva 17-1440-02, pack of 6 × 100 mL
Sigma-Aldrich
HEPES solution, 1 M, pH 7.0-7.6, sterile-filtered, BioReagent, suitable for cell culture