Merck
  • Circ-ZEB1.33 promotes the proliferation of human HCC by sponging miR-200a-3p and upregulating CDK6.
Circ-ZEB1.33 promotes the proliferation of human HCC by sponging miR-200a-3p and upregulating CDK6.
Cancer cell international (2018-08-21)
Yuhua Gong, Jinzhong Mao, Di Wu, Xuemei Wang, Long Li, Liang Zhu, Rong Song
ABSTRACT
Accumulating data indicated that circRNA plays important roles in regulating many biological processes of the tumor, the present study is designated for exploring roles of the circ-ZEB1.33-miR-200a-3p-CDK6 regulating axis in human hepatocellular carcinoma (HCC). The regulation axis as predicted by using online tool circNet, the expression and correlation of circ-ZEB1.33-miR-200a-3p-CDK6 was verified in human HCC. The diagnostic value of both tumor and serum circ-ZEB1.33 was estimated by using clinical samples. The roles of circ-ZEB1.33-miR-200a-3p-CDK6 in regulating cell cycle were explored by using in vitro studies. Overexpression of circ-ZEB1.33 and CDK6, downregulation of miR-200a-3p were detected in human HCC tissues, negative correlation between circ-ZEB1.33 and miR-200a-3p, positive correlation between circ-ZEB1.33 and CDK6 were confirmed in human HCC tissues. Tissue and serum circ-ZEB1.33 were related to different TMN stages and prognosis in HCC patients. RNA pull-down assay implied that circ-ZEB1.33 could decrease miR-200a-3p by sponging miR-200a-3p, and the luciferase reporter assay indicated that miR-200a-3p could downregulate CDK6 transcription by targeting its 3'UTR. The in vitro assays indicated that circ-ZEB1.33 could promote the proliferation of HCC cells by increasing the percentage of S phase regulated by CDK6/Rb. Proliferation promotion roles of the circ-ZEB1.33-miR-200a-3p-CDK6 regulating axis are existed and verified in human HCC, both tumor and serum circ-ZEB1.33 can serve as an indicator for the prognosis of HCC patients.
Sigma-Aldrich
GenElute Plasma/Serum RNA Purification Mini Kit