Merck
  • Efficacy and safety assessment of a TRAF6-targeted nanoimmunotherapy in atherosclerotic mice and non-human primates.
Efficacy and safety assessment of a TRAF6-targeted nanoimmunotherapy in atherosclerotic mice and non-human primates.
Nature biomedical engineering (2019-04-03)
Marnix Lameijer, Tina Binderup, Mandy M T van Leent, Max L Senders, Francois Fay, Joost Malkus, Brenda L Sanchez-Gaytan, Abraham J P Teunissen, Nicolas Karakatsanis, Philip Robson, Xianxiao Zhou, Yuxiang Ye, Gregory Wojtkiewicz, Jun Tang, Tom T P Seijkens, Jeffrey Kroon, Erik S G Stroes, Andreas Kjaer, Jordi Ochando, Thomas Reiner, Carlos Pérez-Medina, Claudia Calcagno, Edward A Fisher, Bin Zhang, Ryan E Temel, Filip K Swirski, Matthias Nahrendorf, Zahi A Fayad, Esther Lutgens, Willem J M Mulder, Raphaël Duivenvoorden
ABSTRACT
Macrophage accumulation in atherosclerosis is directly linked to the destabilization and rupture of plaque, causing acute atherothrombotic events. Circulating monocytes enter the plaque and differentiate into macrophages, where they are activated by CD4+ T lymphocytes through CD40-CD40 ligand signalling. Here, we report the development and multiparametric evaluation of a nanoimmunotherapy that moderates CD40-CD40 ligand signalling in monocytes and macrophages by blocking the interaction between CD40 and tumour necrosis factor receptor-associated factor 6 (TRAF6). We evaluated the biodistribution characteristics of the nanoimmunotherapy in apolipoprotein E-deficient (Apoe-/-) mice and in non-human primates by in vivo positron-emission tomography imaging. In Apoe-/- mice, a 1-week nanoimmunotherapy treatment regimen achieved significant anti-inflammatory effects, which was due to the impaired migration capacity of monocytes, as established by a transcriptome analysis. The rapid reduction of plaque inflammation by the TRAF6-targeted nanoimmunotherapy and its favourable toxicity profiles in both mice and non-human primates highlights the translational potential of this strategy for the treatment of atherosclerosis.
Product Description
Millipore
MILLIPLEX® Mouse Cytokine/Chemokine Magnetic Bead Panel - Immunology Multiplex Assay, Simultaneously analyze multiple cytokine and chemokine biomarkers with Bead-Based Multiplex Assays using the Luminex technology, in mouse serum, plasma and cell culture samples.
Millipore
MILLIPLEX® Mouse Acute Phase Magnetic Bead Panel 2 - Cardiovascular Disease Multiplex Assay, The analytes available for this multiplex kit are: Adipsin, Alpha-1 Acid Glycoprotein (AGP), Alpha-2 Macroglobulin (A2M), C-Reactive Protein, Haptoglobin, SAP.