Merck
Reversible effects of photodamage directed toward mitochondria.
Photochemistry and photobiology (2014-04-26)
David Kessel
ABSTRACT
When the initial effect of photodynamic therapy (PDT) involves mitochondrial photodamage, an early effect is loss of the mitochondrial membrane potential (ΔΨm ). Using murine hepatoma 1c1c7 cells and a photosensitizing agent known to target mitochondria, we examined loss of ΔΨm , initiation of apoptosis and loss of viability as a function of time and light dose. There was a correlation between loss of viability and the rapid disappearance of ΔΨm, as detected by the potential-sensitive probe Mitotracker Orange (MTO). Loss of ΔΨm was, however, reversible even with a substantial loss of viability. Unless there was a supralethal level of photodamage, 1c1c7 cells recovered their mitochondrial membrane potential, even if the cell population was on the pathway to apoptosis and cell death. These results indicate that when mitochondria are the initial PDT target, a qualitative estimate of photokilling can be provided by assessing the initial loss of ΔΨm.
Sigma-Aldrich
Total Protein Kit, Micro-Lowry, Onishi & Barr Modification, sufficient for ~50 manual assays